ENA183888904 Enamine

5-(1-naphthyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09812391
  • Cas Number : 1016494-31-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184...186
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39994

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39994
Availability details for
Quantity
  • Availability
  • Add to Cart