ENA183888907 Enamine

2-[(2-methoxyphenyl)amino]thieno[2,3-d][1,3]thiazole-5-carboxylic acid

  • MDL number MFCD09728879
  • Cas Number : 953726-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39999
Availability details for
Quantity
  • Availability
  • Add to Cart