ENA183888908 Enamine

2-(azepan-1-yl)propan-1-amine

  • MDL number MFCD06446942
  • Cas Number : 900718-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40000
Availability details for
Quantity
  • Availability
  • Add to Cart