ENA183888912 Enamine

1-(2,5-difluorophenyl)ethan-1-ol

  • MDL number MFCD08442989
  • Cas Number : 3874-31-5
  • Molecular Weight : 158.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-40005

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40005
Availability details for
Quantity
  • Availability
  • Add to Cart