ENA183888915 Enamine

1-(3-phenoxyphenyl)methanamine

  • MDL number MFCD01631862
  • Cas Number : 50742-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40013

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40013
Availability details for
Quantity
  • Availability
  • Add to Cart