ENA183888916 Enamine

1-(3,4-dichlorobenzoyl)piperazine

  • MDL number MFCD08444694
  • Cas Number : 477204-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40014

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40014
Availability details for
Quantity
  • Availability
  • Add to Cart