ENA183888920 Enamine

4-amino-N-methyl-N-(prop-2-en-1-yl)benzene-1-sulfonamide

  • MDL number MFCD11505548
  • Cas Number : 848652-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-40019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40019
Availability details for
Quantity
  • Availability
  • Add to Cart