ENA183888930 Enamine

2-(6-nitro-1H-indazol-1-yl)acetic acid

  • MDL number MFCD09805059
  • Cas Number : 857801-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40032
Availability details for
Quantity
  • Availability
  • Add to Cart