ENA183888948 Enamine

3-[(3-bromophenyl)methanesulfonyl]propanoic acid

  • MDL number MFCD09941790
  • Cas Number : 1017090-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40083
Availability details for
Quantity
  • Availability
  • Add to Cart