ENA183888954 Enamine

3-amino-N-propylbenzamide

  • MDL number MFCD09044166
  • Cas Number : 81882-78-2
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-40093

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40093
Availability details for
Quantity
  • Availability
  • Add to Cart