ENA183888961 Enamine

cyclohexanecarbothioamide

  • MDL number MFCD06191625
  • Cas Number : 7390-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40100
Availability details for
Quantity
  • Availability
  • Add to Cart