ENA183888972 Enamine

1-benzyl-5-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD07790148
  • Cas Number : 288252-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40120
Availability details for
Quantity
  • Availability
  • Add to Cart