ENA183888975 Enamine

{4-[(3-methylpiperidin-1-yl)methyl]phenyl}methanamine

  • MDL number MFCD08444579
  • Cas Number : 923137-50-2
  • Molecular Weight : 218.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-40123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40123
Availability details for
Quantity
  • Availability
  • Add to Cart