ENA183888983 Enamine

2-(1H-imidazol-1-yl)propanoic acid hydrochloride

  • MDL number MFCD06800800
  • Cas Number : 90269-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40143
Availability details for
Quantity
  • Availability
  • Add to Cart