ENA183888993 Enamine

4-ethoxybutanoic acid

  • MDL number MFCD09928843
  • Cas Number : 10374-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40154
Availability details for
Quantity
  • Availability
  • Add to Cart