ENA183889006 Enamine

(3-fluoro-4-methylphenyl)urea

  • MDL number MFCD09727592
  • Cas Number : 953736-14-6
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40183
Availability details for
Quantity
  • Availability
  • Add to Cart