ENA183889011 Enamine

4-(3-bromophenoxy)pyridine

  • MDL number MFCD09931381
  • Cas Number : 1016494-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40188

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40188
Availability details for
Quantity
  • Availability
  • Add to Cart