ENA183889014 Enamine

2-[1-(3-chloro-4-methylphenyl)-3,5-dimethyl-1H-pyrazol-4-yl]acetic acid

  • MDL number MFCD09044278
  • Cas Number : 926265-54-5
  • Molecular Weight : 278.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [1-(3-chloro-4-methylphenyl)-3,5-dimethyl-1H-pyrazol-4-yl]acetic acid
Inchi Code 1S/C14H15ClN2O2/c1-8-4-5-11(6-13(8)15)17-10(3)12(7-14(18)19)9(2)16-17/h4-6H,7H2,1-3H3,(H,18,19)
Inchi Key BGPVEEYVOYVYOZ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-40193

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40193
Availability details for
Quantity
  • Availability
  • Add to Cart