ENA183889015 Enamine

2-[1-(2-fluorophenyl)-3,5-dimethyl-1H-pyrazol-4-yl]acetic acid

  • MDL number MFCD09042766
  • Cas Number : 926211-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40195
Availability details for
Quantity
  • Availability
  • Add to Cart