ENA183889020 Enamine

2-(3-methylpiperidin-1-yl)propan-1-amine

  • MDL number MFCD06446937
  • Cas Number : 900717-97-7
  • Molecular Weight : 156.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40202
Availability details for
Quantity
  • Availability
  • Add to Cart