ENA183889027 Enamine

8-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carboxylic acid

  • MDL number MFCD00067818
  • Cas Number : 34662-59-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-40213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40213
Availability details for
Quantity
  • Availability
  • Add to Cart