ENA183889028 Enamine

tert-butyl[(3-fluorophenyl)methyl]amine

  • MDL number MFCD09044475
  • Cas Number : 926198-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-40214

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40214
Availability details for
Quantity
  • Availability
  • Add to Cart