ENA183889029 Enamine

2-{3,5-dimethyl-1-[3-(trifluoromethyl)phenyl]-1H-pyrazol-4-yl}acetic acid

  • MDL number MFCD09046265
  • Cas Number : 70597-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40216
Availability details for
Quantity
  • Availability
  • Add to Cart