ENA183889039 Enamine

2-[2-(2-hydroxy-5-methylbenzamido)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD09813671
  • Cas Number : 1016496-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-40226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40226
Availability details for
Quantity
  • Availability
  • Add to Cart