ENA183889049 Enamine

N-(pyridin-3-ylmethyl)urea

  • MDL number MFCD00134234
  • Cas Number : 36226-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182...184
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40239

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40239
Availability details for
Quantity
  • Availability
  • Add to Cart