ENA183889062 Enamine

2-methyl-5H,6H,7H,8H-pyrido[4,3-d]pyrimidine

  • MDL number MFCD09040774
  • Cas Number : 676994-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40280
Availability details for
Quantity
  • Availability
  • Add to Cart