ENA183889065 Enamine

(2Z)-2-(2,4-dimethoxybenzylidene)-6-hydroxy-1-benzofuran-3(2H)-one

  • MDL number MFCD04065081
  • Cas Number : 210360-63-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36353

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36353
Availability details for
Quantity
  • Availability
  • Add to Cart