ENA183889067 Enamine

N-[4-amino-3-(trifluoromethyl)phenyl]benzamide

  • MDL number MFCD09044387
  • Cas Number : 736-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-40287

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40287
Availability details for
Quantity
  • Availability
  • Add to Cart