ENA183889073 Enamine

(2Z)-2-(2-ethoxybenzylidene)-6-hydroxy-1-benzofuran-3(2H)-one

  • MDL number MFCD04065082
  • Cas Number : 623122-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36356

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36356
Availability details for
Quantity
  • Availability
  • Add to Cart