ENA183889081 Enamine

N-(3-amino-4-methylphenyl)-2-methylpropanamide

  • MDL number MFCD09932408
  • Cas Number : 946690-30-8
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40308
Availability details for
Quantity
  • Availability
  • Add to Cart