ENA183889094 Enamine

N-[4-amino-3-(trifluoromethyl)phenyl]-4-fluorobenzamide

  • MDL number MFCD09045200
  • Cas Number : 926253-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40324

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40324
Availability details for
Quantity
  • Availability
  • Add to Cart