ENA183889099 Enamine

3-(1H-1,3-benzodiazol-1-yl)-N'-hydroxypropanimidamide

  • Cas Number : 4404-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40331

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40331
Availability details for
Quantity
  • Availability
  • Add to Cart