ENA183889101 Enamine

3-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)propanamide

  • MDL number MFCD00833539
  • Cas Number : 116857-94-4
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-40333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40333
Availability details for
Quantity
  • Availability
  • Add to Cart