ENA183889106 Enamine

2-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-N'-hydroxyethanimidamide

  • MDL number MFCD08436508
  • Cas Number : 889951-80-8
  • Molecular Weight : 218.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-40339

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40339
Availability details for
Quantity
  • Availability
  • Add to Cart