ENA183889114 Enamine

[(pyridin-2-yl)methyl]urea

  • MDL number MFCD00276112
  • Cas Number : 36226-31-0
  • Molecular Weight : 151.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-40360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40360
Availability details for
Quantity
  • Availability
  • Add to Cart