ENA183889127 Enamine

ethyl 2-amino-3-oxobutanoate hydrochloride

  • MDL number MFCD09971784
  • Cas Number : 20207-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-123
Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40378
Availability details for
Quantity
  • Availability
  • Add to Cart