ENA183889132 Enamine

[4-(4-methylphenyl)-1,3-thiazol-2-yl]methanamine

  • MDL number MFCD05221841
  • Cas Number : 643723-47-1
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40389
Availability details for
Quantity
  • Availability
  • Add to Cart