ENA183889135 Enamine

cyclopentanecarboxamide

  • MDL number MFCD11505594
  • Cas Number : 3217-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40396

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40396
Availability details for
Quantity
  • Availability
  • Add to Cart