ENA183889143 Enamine

2-(3-methyl-2,5-dioxoimidazolidin-1-yl)-N-phenylacetamide

  • MDL number MFCD03972501
  • Cas Number : 730253-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 279...281
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40405

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40405
Availability details for
Quantity
  • Availability
  • Add to Cart