ENA183889149 Enamine

4-[(2-fluorophenyl)sulfamoyl]-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD09052655
  • Cas Number : 937621-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40414
Availability details for
Quantity
  • Availability
  • Add to Cart