ENA183889151 Enamine

carbamothioylmethyl acetate

  • MDL number MFCD11505599
  • Cas Number : 40235-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40416
Availability details for
Quantity
  • Availability
  • Add to Cart