ENA183889153 Enamine

N-(3-aminopropyl)-N-methyl-N-(2,2,2-trifluoroethyl)amine

  • MDL number MFCD11130122
  • Cas Number : 1019397-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40418

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40418
Availability details for
Quantity
  • Availability
  • Add to Cart