ENA183889157 Enamine

4-[(3-methylpiperidin-1-yl)methyl]aniline

  • MDL number MFCD09036983
  • Cas Number : 926207-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40423
Availability details for
Quantity
  • Availability
  • Add to Cart