ENA183889169 Enamine

6-(benzyloxy)pyridine-3-carboxylic acid

  • MDL number MFCD09730715
  • Cas Number : 94084-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-40450

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40450
Availability details for
Quantity
  • Availability
  • Add to Cart