ENA183889186 Enamine

5-(chlorosulfonyl)-2,3-dimethoxybenzoic acid

  • MDL number MFCD09048188
  • Cas Number : 74651-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40475
Availability details for
Quantity
  • Availability
  • Add to Cart