ENA183889188 Enamine

4-(benzylsulfanyl)-2-chloropyridine

  • MDL number MFCD11505608
  • Cas Number : 71506-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40477
Availability details for
Quantity
  • Availability
  • Add to Cart