ENA183889189 Enamine

3-methyl-2-(pyrrolidin-1-ylcarbonyl)aniline

  • MDL number MFCD09734975
  • Cas Number : 953746-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87...89
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40488

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40488
Availability details for
Quantity
  • Availability
  • Add to Cart