ENA183889212 Enamine

2-[4-(3,5-dimethyl-1H-pyrazol-1-yl)phenyl]ethan-1-amine

  • MDL number MFCD08443148
  • Cas Number : 616879-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 91
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40520
Availability details for
Quantity
  • Availability
  • Add to Cart