ENA183889214 Enamine

4-(aminomethyl)-N-ethylbenzamide

  • MDL number MFCD09050879
  • Cas Number : 777055-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-40522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40522
Availability details for
Quantity
  • Availability
  • Add to Cart