ENA183889228 Enamine

1-[4-(piperidin-1-ylcarbonyl)phenyl]methanamine

  • MDL number MFCD07366441
  • Cas Number : 292635-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40754

Safety Information

msds https://www.enaminestore.com/catalog/EN300-40754
Availability details for
Quantity
  • Availability
  • Add to Cart